Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGCGTCGGTTGGGACGCGGAGC[C/T]GAGGAGCAGGGCCGGGCGCCATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHPP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LHPP - phospholysine phosphohistidine inorganic pyrophosphate phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167880.1 | 71 | UTR 5 | NP_001161352.1 | |||
NM_001318331.1 | 71 | UTR 5 | NP_001305260.1 | |||
NM_001318332.1 | 71 | UTR 5 | NP_001305261.1 | |||
NM_022126.3 | 71 | UTR 5 | NP_071409.3 | |||
XM_005270026.3 | 71 | UTR 5 | XP_005270083.1 | |||
XM_006717943.1 | 71 | UTR 5 | XP_006718006.1 | |||
XM_011540058.2 | 71 | UTR 5 | XP_011538360.1 | |||
XM_017016509.1 | 71 | UTR 5 | XP_016871998.1 | |||
XM_017016510.1 | 71 | UTR 5 | XP_016871999.1 | |||
XM_017016511.1 | 71 | UTR 5 | XP_016872000.1 | |||
XM_017016512.1 | 71 | Intron | XP_016872001.1 |