Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTCACACGGAATGGCATATCGA[A/T]CAATCTTGTATTGGAATGACCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKIB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKIB1 - ankyrin repeat and IBR domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019004.1 | Intron | NP_061877.1 | ||||
XM_005250458.3 | Intron | XP_005250515.1 | ||||
XM_011516334.1 | Intron | XP_011514636.1 | ||||
XM_011516335.2 | Intron | XP_011514637.1 | ||||
XM_017012359.1 | Intron | XP_016867848.1 | ||||
XM_017012360.1 | Intron | XP_016867849.1 |
KRIT1 - KRIT1, ankyrin repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |