Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGACTGCTGGAAGAATTAATTAG[A/G]AACGTGCTGTGCTCTGGGCAGCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NTNG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NTNG2 - netrin G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032536.2 | Intron | NP_115925.2 | ||||
XM_006717304.3 | Intron | XP_006717367.1 | ||||
XM_011519094.2 | Intron | XP_011517396.1 | ||||
XM_011519096.2 | Intron | XP_011517398.1 | ||||
XM_011519097.2 | Intron | XP_011517399.1 | ||||
XM_011519098.2 | Intron | XP_011517400.1 | ||||
XM_011519099.2 | Intron | XP_011517401.1 | ||||
XM_011519100.2 | Intron | XP_011517402.1 | ||||
XM_011519102.2 | Intron | XP_011517404.1 | ||||
XM_011519103.2 | Intron | XP_011517405.1 | ||||
XM_011519104.2 | Intron | XP_011517406.1 | ||||
XM_011519105.2 | Intron | XP_011517407.1 | ||||
XM_011519106.2 | Intron | XP_011517408.1 | ||||
XM_011519107.2 | Intron | XP_011517409.1 | ||||
XM_011519108.2 | Intron | XP_011517410.1 | ||||
XM_011519109.2 | Intron | XP_011517411.1 | ||||
XM_011519110.2 | Intron | XP_011517412.1 | ||||
XM_011519112.2 | Intron | XP_011517414.1 | ||||
XM_011519113.2 | Intron | XP_011517415.1 | ||||
XM_017015212.1 | Intron | XP_016870701.1 | ||||
XM_017015213.1 | Intron | XP_016870702.1 | ||||
XM_017015214.1 | Intron | XP_016870703.1 | ||||
XM_017015215.1 | Intron | XP_016870704.1 | ||||
XM_017015216.1 | Intron | XP_016870705.1 |