Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGGAGCTGCGGACTCACTGGAGC[C/G]GAGAACTGGGGCGGCGCGGCGCGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616692 | |||||||||||||||||||||||
Literature Links: |
ESYT3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ESYT3 - extended synaptotagmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322831.1 | 13 | UTR 5 | NP_001309760.1 | |||
NM_001322834.1 | 13 | UTR 5 | NP_001309763.1 | |||
NM_031913.4 | 13 | UTR 5 | NP_114119.2 | |||
XM_017007307.1 | 13 | Intron | XP_016862796.1 |