Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCCACAGGATTGCCTTCCATCT[C/G]CTCTTGGTCCCTCCTGGATGTGGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120470 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCC - DCC netrin 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005215.3 | 242 | UTR 5 | NP_005206.2 | |||
XM_011525843.1 | 242 | UTR 5 | XP_011524145.1 | |||
XM_011525844.2 | 242 | Intron | XP_011524146.1 | |||
XM_017025568.1 | 242 | UTR 5 | XP_016881057.1 | |||
XM_017025569.1 | 242 | UTR 5 | XP_016881058.1 | |||
XM_017025570.1 | 242 | Intron | XP_016881059.1 |