Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGCTCACATTTGGATCTAACC[G/T]GACTCCAGAATGCCTATGAACACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609171 MIM: 616575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42EP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC42EP5 - CDC42 effector protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145057.3 | Intron | NP_659494.2 |
LENG8 - leukocyte receptor cluster member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LENG9 - leukocyte receptor cluster member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |