Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTTTAGATGGAGGATTGTTCTG[A/C]TTGTTCTGATTAGTCCACAGTTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616912 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EVL - Enah/Vasp-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016337.2 | Intron | NP_057421.1 | ||||
XM_005267749.3 | Intron | XP_005267806.1 | ||||
XM_011536828.2 | Intron | XP_011535130.1 | ||||
XM_017021363.1 | Intron | XP_016876852.1 | ||||
XM_017021364.1 | Intron | XP_016876853.1 |