Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGGGAGTGCTTCCTCGCTCTC[A/C]GAGTGCGCAAGCGCAGCGCACCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601505 MIM: 614275 | ||||||||||||||||||||
Literature Links: |
ZNF146 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF146 - zinc finger protein 146 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099638.1 | 504 | UTR 5 | NP_001093108.1 | |||
NM_001099639.1 | 504 | UTR 5 | NP_001093109.1 | |||
NM_007145.2 | 504 | UTR 5 | NP_009076.2 | |||
XM_005259214.2 | 504 | UTR 5 | XP_005259271.1 | |||
XM_017027244.1 | 504 | Intron | XP_016882733.1 | |||
XM_017027245.1 | 504 | Intron | XP_016882734.1 | |||
XM_017027246.1 | 504 | Intron | XP_016882735.1 | |||
XM_017027247.1 | 504 | Intron | XP_016882736.1 |
ZNF565 - zinc finger protein 565 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042474.2 | 504 | Intron | NP_001035939.1 | |||
NM_152477.4 | 504 | Intron | NP_689690.3 | |||
XM_011526512.2 | 504 | Intron | XP_011524814.1 | |||
XM_011526514.2 | 504 | Intron | XP_011524816.1 | |||
XM_017026341.1 | 504 | Intron | XP_016881830.1 | |||
XM_017026342.1 | 504 | Intron | XP_016881831.1 | |||
XM_017026343.1 | 504 | Intron | XP_016881832.1 |