Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTTGTGCCTTGAAGCAGAGGG[C/T]GGGAGCAGCTGGTAAGATGCAGCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613766 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCAMP5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SCAMP5 - secretory carrier membrane protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178111.1 | Intron | NP_001171582.1 | ||||
NM_001178112.1 | Intron | NP_001171583.1 | ||||
NM_138967.3 | Intron | NP_620417.1 | ||||
XM_006720420.1 | Intron | XP_006720483.1 | ||||
XM_006720421.1 | Intron | XP_006720484.1 | ||||
XM_006720422.3 | Intron | XP_006720485.1 | ||||
XM_017021989.1 | Intron | XP_016877478.1 |
Set Membership: |
HapMap |