Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCGGGGCGAGTGGGGAACCCGG[A/C]GCAGGAACTGCAGCCGCGGCTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180201 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID4A - AT-rich interaction domain 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002892.3 | 208 | UTR 5 | NP_002883.3 | |||
NM_023000.2 | 208 | UTR 5 | NP_075376.2 | |||
NM_023001.2 | 208 | UTR 5 | NP_075377.2 | |||
XM_005267964.2 | 208 | UTR 5 | XP_005268021.1 | |||
XM_005267965.2 | 208 | UTR 5 | XP_005268022.1 | |||
XM_017021560.1 | 208 | UTR 5 | XP_016877049.1 | |||
XM_017021561.1 | 208 | Intron | XP_016877050.1 | |||
XM_017021562.1 | 208 | UTR 5 | XP_016877051.1 | |||
XM_017021563.1 | 208 | UTR 5 | XP_016877052.1 | |||
XM_017021564.1 | 208 | Intron | XP_016877053.1 | |||
XM_017021565.1 | 208 | Intron | XP_016877054.1 |
PSMA3-AS1 - PSMA3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |