Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAAACCCACCTCCCCTCTCCCC[C/T]TGGGCAATTTTTATCTCCCTTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHD3 - chromodomain helicase DNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005271.2 | 133 | Intron | NP_001005271.2 | |||
NM_001005273.2 | 133 | Intron | NP_001005273.1 | |||
NM_005852.3 | 133 | Intron | NP_005843.2 | |||
XM_005256427.4 | 133 | Intron | XP_005256484.1 | |||
XM_005256428.4 | 133 | Intron | XP_005256485.1 | |||
XM_005256429.4 | 133 | Intron | XP_005256486.1 | |||
XM_005256431.4 | 133 | Intron | XP_005256488.1 | |||
XM_006721423.3 | 133 | Intron | XP_006721486.1 | |||
XM_006721424.3 | 133 | Intron | XP_006721487.1 | |||
XM_006721428.3 | 133 | Intron | XP_006721491.1 | |||
XM_017024061.1 | 133 | Intron | XP_016879550.1 | |||
XM_017024062.1 | 133 | Intron | XP_016879551.1 | |||
XM_017024063.1 | 133 | Intron | XP_016879552.1 | |||
XM_017024064.1 | 133 | Intron | XP_016879553.1 | |||
XM_017024065.1 | 133 | Intron | XP_016879554.1 | |||
XM_017024066.1 | 133 | Intron | XP_016879555.1 | |||
XM_017024067.1 | 133 | UTR 5 | XP_016879556.1 | |||
XM_017024068.1 | 133 | UTR 5 | XP_016879557.1 | |||
XM_017024069.1 | 133 | Intron | XP_016879558.1 | |||
XM_017024070.1 | 133 | Intron | XP_016879559.1 |
NAA38 - N(alpha)-acetyltransferase 38, NatC auxiliary subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |