Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCATGTGTACACATGTCAGAGG[C/T]CCCAGACATGTCAACTTTTCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614440 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PSD3 - pleckstrin and Sec7 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015310.3 | 10826 | UTR 3 | NP_056125.3 | |||
NM_206909.2 | 10826 | UTR 3 | NP_996792.1 | |||
XM_011544467.2 | 10826 | Intron | XP_011542769.1 | |||
XM_011544468.2 | 10826 | Intron | XP_011542770.1 | |||
XM_011544469.2 | 10826 | Intron | XP_011542771.1 | |||
XM_011544471.2 | 10826 | Intron | XP_011542773.1 | |||
XM_011544473.2 | 10826 | Intron | XP_011542775.1 | |||
XM_017013258.1 | 10826 | UTR 3 | XP_016868747.1 | |||
XM_017013259.1 | 10826 | UTR 3 | XP_016868748.1 | |||
XM_017013260.1 | 10826 | UTR 3 | XP_016868749.1 | |||
XM_017013261.1 | 10826 | UTR 3 | XP_016868750.1 | |||
XM_017013262.1 | 10826 | Intron | XP_016868751.1 | |||
XM_017013263.1 | 10826 | Intron | XP_016868752.1 | |||
XM_017013264.1 | 10826 | Intron | XP_016868753.1 | |||
XM_017013265.1 | 10826 | UTR 3 | XP_016868754.1 |
Set Membership: |
HapMap |