Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCATCTTTGGAAGAAAACCCACA[C/T]AGTTTTTTCAGGAGCTGAAAATTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID5B - AT-rich interaction domain 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244638.1 | 240 | Intron | NP_001231567.1 | |||
NM_032199.2 | 240 | UTR 5 | NP_115575.1 | |||
XM_011540262.1 | 240 | UTR 5 | XP_011538564.1 | |||
XM_017016765.1 | 240 | Intron | XP_016872254.1 |