Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTAGGGACCTCCTAGAGGACCA[C/T]GTGCAGGTGGGGCCAGGGAGGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606268 MIM: 604663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNT10A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WNT10A - Wnt family member 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025216.2 | Intron | NP_079492.2 | ||||
XM_011511929.2 | Intron | XP_011510231.1 | ||||
XM_011511930.1 | Intron | XP_011510232.1 |
WNT6 - Wnt family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |