Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTTACCAGCAGATGCATACAC[A/G]TAATTGGGTACTTCATTGCTAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKMY2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKMY2 - ankyrin repeat and MYND domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BZW2 - basic leucine zipper and W2 domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159767.1 | Intron | NP_001153239.1 | ||||
NM_014038.2 | Intron | NP_054757.1 | ||||
XM_006715706.1 | Intron | XP_006715769.1 | ||||
XM_006715707.1 | Intron | XP_006715770.1 | ||||
XM_006715708.1 | Intron | XP_006715771.1 |