Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCTGGGAGAGGATGATCCATAG[A/G]CAATGAGTGTCTCTAAGAGGGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPIN3 - spindlin family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010862.2 | 215 | UTR 5 | NP_001010862.2 | |||
XM_006724577.3 | 215 | UTR 5 | XP_006724640.1 | |||
XM_006724578.3 | 215 | UTR 5 | XP_006724641.1 | |||
XM_017029315.1 | 215 | UTR 5 | XP_016884804.1 |