Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGCGGCCGAAAACGCCAAGAGA[G/T]TGATGCTGTAGCTGCCCTGAGATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611661 | ||||||||||||||||||||
Literature Links: |
DBF4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DBF4B - DBF4 zinc finger B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025104.3 | 63 | UTR 5 | NP_079380.2 | |||
NM_145663.2 | 63 | UTR 5 | NP_663696.1 | |||
XM_006722098.2 | 63 | UTR 5 | XP_006722161.1 | |||
XM_006722102.3 | 63 | UTR 5 | XP_006722165.1 | |||
XM_011525281.2 | 63 | Intron | XP_011523583.1 | |||
XM_017025145.1 | 63 | UTR 5 | XP_016880634.1 | |||
XM_017025146.1 | 63 | Intron | XP_016880635.1 | |||
XM_017025147.1 | 63 | UTR 5 | XP_016880636.1 | |||
XM_017025148.1 | 63 | UTR 5 | XP_016880637.1 | |||
XM_017025149.1 | 63 | UTR 5 | XP_016880638.1 | |||
XM_017025150.1 | 63 | UTR 5 | XP_016880639.1 | |||
XM_017025151.1 | 63 | UTR 5 | XP_016880640.1 | |||
XM_017025152.1 | 63 | Intron | XP_016880641.1 | |||
XM_017025153.1 | 63 | Intron | XP_016880642.1 | |||
XM_017025154.1 | 63 | Intron | XP_016880643.1 | |||
XM_017025155.1 | 63 | UTR 5 | XP_016880644.1 | |||
XM_017025156.1 | 63 | UTR 5 | XP_016880645.1 |