Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAAAGGCAGTAAGATGATAACC[C/T]ATACTATGAACCATAAAACAACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SULT1C3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SULT1C3 - sulfotransferase family 1C member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008743.2 | 917 | Intron | NP_001008743.1 | |||
NM_001320878.1 | 917 | Intron | NP_001307807.1 | |||
XM_017004153.1 | 917 | Missense Mutation | CAT,TAT | H,Y 22 | XP_016859642.1 | |
XM_017004154.1 | 917 | Missense Mutation | CAT,TAT | H,Y 22 | XP_016859643.1 | |
XM_017004155.1 | 917 | Missense Mutation | CAT,TAT | H,Y 22 | XP_016859644.1 |