Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTCTCCATTCGAGCACCTTCCAG[A/C]ATACCGCTCGGCTCCGGGAGCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAMDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAMDC2 - MAM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153267.4 | 492 | UTR 5 | NP_694999.3 | |||
XM_011518471.1 | 492 | UTR 5 | XP_011516773.1 | |||
XM_011518475.2 | 492 | UTR 5 | XP_011516777.1 | |||
XM_017014587.1 | 492 | UTR 5 | XP_016870076.1 |