Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGAAGAGAGGCCCAAGATGGAG[A/G]CGGCGGCGGCTGTAGCGGCGTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 311010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARAF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARAF - A-Raf proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256196.1 | 126 | UTR 5 | NP_001243125.1 | |||
NM_001256197.1 | 126 | UTR 5 | NP_001243126.1 | |||
NM_001654.4 | 126 | UTR 5 | NP_001645.1 | |||
XM_006724529.2 | 126 | UTR 5 | XP_006724592.1 | |||
XM_011543906.2 | 126 | Intron | XP_011542208.1 | |||
XM_011543908.2 | 126 | Intron | XP_011542210.1 | |||
XM_011543909.2 | 126 | Intron | XP_011542211.1 |