Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGGTACACAGCGTCCTGAACTG[A/G]GCTATAGAGAGGTCAGGCAAACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLAC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLAC1 - placenta specific 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316887.1 | Intron | NP_001303816.1 | ||||
NM_001316888.1 | Intron | NP_001303817.1 | ||||
NM_001316889.1 | Intron | NP_001303818.1 | ||||
NM_021796.3 | Intron | NP_068568.1 | ||||
XM_011531257.2 | Intron | XP_011529559.1 | ||||
XM_011531260.2 | Intron | XP_011529562.1 | ||||
XM_017029236.1 | Intron | XP_016884725.1 |