Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGTTTCTGTAACCTCCTGACA[C/T]GGACGGCGACTCTGGTCTCGCAGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606765 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPO PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TPO - thyroid peroxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000547.5 | Intron | NP_000538.3 | ||||
NM_001206744.1 | Intron | NP_001193673.1 | ||||
NM_001206745.1 | Intron | NP_001193674.1 | ||||
NM_175719.3 | Intron | NP_783650.1 | ||||
NM_175721.3 | Intron | NP_783652.1 | ||||
NM_175722.3 | Intron | NP_783653.1 | ||||
XM_011510379.2 | Intron | XP_011508681.1 | ||||
XM_011510380.2 | Intron | XP_011508682.1 | ||||
XM_011510381.2 | Intron | XP_011508683.1 |
Set Membership: |
HapMap |