Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGAGGAGCAGGGACTTACAGAG[C/T]TCCTCGGGGTCAGACAGTTGGTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607875 MIM: 600174 | ||||||||||||||||||||
Literature Links: |
INPP5K PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INPP5K - inositol polyphosphate-5-phosphatase K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNA - phosphatidylinositol transfer protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006224.3 | 1911 | UTR 3 | NP_006215.1 |
PITPNA-AS1 - PITPNA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |