Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGATTCTGCCCTTGACCCTGTT[A/G]AGGCACGGCAGATGGTCTTCATCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF879 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF879 - zinc finger protein 879 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136116.1 | 659 | Intron | NP_001129588.1 | |||
XM_005265908.3 | 659 | UTR 5 | XP_005265965.1 | |||
XM_011534550.2 | 659 | Intron | XP_011532852.1 | |||
XM_011534551.2 | 659 | Intron | XP_011532853.1 |