Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607639 MIM: 182889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSC4D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SSC4D - scavenger receptor cysteine rich family member with 4 domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080744.1 | 731 | Missense Mutation | CAC,CGC | H,R 128 | NP_542782.1 | |
XM_017011750.1 | 731 | Intron | XP_016867239.1 |
ZP3 - zona pellucida glycoprotein 3 (sperm receptor) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110354.1 | 731 | Intron | NP_001103824.1 | |||
NM_007155.5 | 731 | Intron | NP_009086.4 | |||
XM_017012586.1 | 731 | Intron | XP_016868075.1 |
Set Membership: |
HapMap |