Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAGAACACCAGCGCCCTCCCTC[C/T]GGTGCAGCCCTGCCTGGCCGGGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603418 MIM: 614760 | ||||||||||||||||||||
Literature Links: |
AKR7A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKR7A2 - aldo-keto reductase family 7 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PQLC2 - PQ loop repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040125.1 | 682 | UTR 5 | NP_001035214.1 | |||
NM_001040126.1 | 682 | Intron | NP_001035215.1 | |||
NM_001287531.1 | 682 | Intron | NP_001274460.1 | |||
NM_017765.2 | 682 | UTR 5 | NP_060235.2 | |||
XM_005245916.2 | 682 | UTR 5 | XP_005245973.1 | |||
XM_006710710.2 | 682 | UTR 5 | XP_006710773.1 | |||
XM_006710711.2 | 682 | UTR 5 | XP_006710774.1 | |||
XM_011541634.2 | 682 | UTR 5 | XP_011539936.1 |