Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGGCTGTCTCCTCCTCTTCCCC[A/G]GCAGCTGCTGCTCGCTCAGCTCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAMA2 - laminin subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000426.3 | 7 | UTR 5 | NP_000417.2 | |||
NM_001079823.1 | 7 | UTR 5 | NP_001073291.1 | |||
XM_005266981.3 | 7 | UTR 5 | XP_005267038.1 | |||
XM_005266982.3 | 7 | UTR 5 | XP_005267039.1 | |||
XM_011535820.2 | 7 | UTR 5 | XP_011534122.1 | |||
XM_017010851.1 | 7 | Intron | XP_016866340.1 | |||
XM_017010852.1 | 7 | Intron | XP_016866341.1 | |||
XM_017010853.1 | 7 | UTR 5 | XP_016866342.1 |