Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATTGTGTTACACCTTGGATTTG[C/T]GTAATCCCTGGATGGCAGTTAGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606188 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM28 - ADAM metallopeptidase domain 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304351.1 | Intron | NP_001291280.1 | ||||
NM_014265.5 | Intron | NP_055080.2 | ||||
NM_021777.4 | Intron | NP_068547.2 | ||||
XM_005273380.3 | Intron | XP_005273437.1 | ||||
XM_005273382.1 | Intron | XP_005273439.1 | ||||
XM_006716273.3 | Intron | XP_006716336.1 | ||||
XM_006716274.1 | Intron | XP_006716337.1 | ||||
XM_011544367.2 | Intron | XP_011542669.1 | ||||
XM_011544368.2 | Intron | XP_011542670.1 | ||||
XM_011544369.2 | Intron | XP_011542671.1 | ||||
XM_011544370.2 | Intron | XP_011542672.1 | ||||
XM_011544371.2 | Intron | XP_011542673.1 | ||||
XM_017012974.1 | Intron | XP_016868463.1 | ||||
XM_017012975.1 | Intron | XP_016868464.1 | ||||
XM_017012976.1 | Intron | XP_016868465.1 |
LOC101929294 - uncharacterized LOC101929294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |