Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACACCAGCTCCGCCAAGGCGCCC[T/G]CGAGGCGAAGCGGGAGTCGAGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609199 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTSL3 - ADAMTS like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301110.1 | Intron | NP_001288039.1 | ||||
NM_207517.2 | Intron | NP_997400.2 | ||||
XM_011521822.2 | Intron | XP_011520124.1 | ||||
XM_011521823.2 | Intron | XP_011520125.1 | ||||
XM_011521824.2 | Intron | XP_011520126.1 | ||||
XM_011521825.2 | Intron | XP_011520127.1 | ||||
XM_017022434.1 | Intron | XP_016877923.1 | ||||
XM_017022435.1 | Intron | XP_016877924.1 |