Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAATTCAGCAAAGCTCTCCCACT[G/T]CAGATTGGGAGAATCAGGTATTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601685 | ||||||||||||||||||||
Literature Links: |
RPS6KA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPS6KA2 - ribosomal protein S6 kinase A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006932.2 | 4936 | UTR 3 | NP_001006933.2 | |||
NM_001318936.1 | 4936 | UTR 3 | NP_001305865.1 | |||
NM_001318937.1 | 4936 | UTR 3 | NP_001305866.1 | |||
NM_001318938.1 | 4936 | UTR 3 | NP_001305867.1 | |||
NM_021135.5 | 4936 | UTR 3 | NP_066958.2 |