Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTAACTTTCTTAAGTTTAAAGT[A/G]TACTATCTCCTGCAGCTCAATATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601269 MIM: 614405 | ||||||||||||||||||||
Literature Links: |
C1QBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1QBP - complement C1q binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DHX33 - DEAH-box helicase 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199699.1 | 4797 | UTR 3 | NP_001186628.1 | |||
NM_020162.3 | 4797 | UTR 3 | NP_064547.2 | |||
XM_017024877.1 | 4797 | Intron | XP_016880366.1 |
RPAIN - RPA interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |