Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTCATCACGGGAACTGCTGGG[C/G]TATGGAATACAGATGTGGCAGCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607667 MIM: 610869 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTNNA3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CTNNA3 - catenin alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127384.2 | 138 | Intron | NP_001120856.1 | |||
NM_001291133.1 | 138 | Intron | NP_001278062.1 | |||
NM_013266.3 | 138 | Intron | NP_037398.2 | |||
XM_017016151.1 | 138 | Intron | XP_016871640.1 | |||
XM_017016152.1 | 138 | Intron | XP_016871641.1 | |||
XM_017016153.1 | 138 | Intron | XP_016871642.1 | |||
XM_017016154.1 | 138 | Intron | XP_016871643.1 | |||
XM_017016155.1 | 138 | Intron | XP_016871644.1 | |||
XM_017016156.1 | 138 | Intron | XP_016871645.1 | |||
XM_017016157.1 | 138 | Intron | XP_016871646.1 | |||
XM_017016158.1 | 138 | Intron | XP_016871647.1 |
LOC101928961 - uncharacterized LOC101928961 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRTM3 - leucine rich repeat transmembrane neuronal 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178011.4 | 138 | UTR 5 | NP_821079.3 |
Set Membership: |
HapMap |