Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATACATTAAAGTCACGTCAATG[C/T]GGCAAGTGCAGTCCCTGGCACGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACCS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACCS - 1-aminocyclopropane-1-carboxylate synthase homolog (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127219.1 | Intron | NP_001120691.1 | ||||
NM_032592.3 | Intron | NP_115981.1 | ||||
XM_005253170.3 | Intron | XP_005253227.2 | ||||
XM_005253172.2 | Intron | XP_005253229.1 | ||||
XM_006718348.2 | Intron | XP_006718411.1 | ||||
XM_011520406.2 | Intron | XP_011518708.1 | ||||
XM_011520407.1 | Intron | XP_011518709.1 | ||||
XM_011520410.2 | Intron | XP_011518712.1 | ||||
XM_017018421.1 | Intron | XP_016873910.1 |
ACCSL - 1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |