Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACTAAAAAACAAAACAAACAAA[C/T]GAAAAAAAAACACAAAACAAACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113505 MIM: 611468 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BDNF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BDNF - brain derived neurotrophic factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143805.1 | 2402 | UTR 3 | NP_001137277.1 | |||
NM_001143806.1 | 2402 | UTR 3 | NP_001137278.1 | |||
NM_001143807.1 | 2402 | UTR 3 | NP_001137279.1 | |||
NM_001143808.1 | 2402 | UTR 3 | NP_001137280.1 | |||
NM_001143809.1 | 2402 | UTR 3 | NP_001137281.1 | |||
NM_001143810.1 | 2402 | UTR 3 | NP_001137282.1 | |||
NM_001143811.1 | 2402 | UTR 3 | NP_001137283.1 | |||
NM_001143812.1 | 2402 | UTR 3 | NP_001137284.1 | |||
NM_001143813.1 | 2402 | UTR 3 | NP_001137285.1 | |||
NM_001143814.1 | 2402 | UTR 3 | NP_001137286.1 | |||
NM_001143816.1 | 2402 | UTR 3 | NP_001137288.1 | |||
NM_001709.4 | 2402 | UTR 3 | NP_001700.2 | |||
NM_170731.4 | 2402 | UTR 3 | NP_733927.1 | |||
NM_170732.4 | 2402 | UTR 3 | NP_733928.1 | |||
NM_170733.3 | 2402 | UTR 3 | NP_733929.1 | |||
NM_170734.3 | 2402 | UTR 3 | NP_733930.1 | |||
NM_170735.5 | 2402 | UTR 3 | NP_733931.1 | |||
XM_011520280.2 | 2402 | UTR 3 | XP_011518582.1 |
BDNF-AS - BDNF antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |