Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATCTTCCACTGTGTGACTCGGG[A/T]AATGGCTGTTCGTGTTTATGATTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601081 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCD2 - ATP binding cassette subfamily D member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005164.3 | Intron | NP_005155.1 | ||||
XM_011538027.2 | Intron | XP_011536329.1 | ||||
XM_017018992.1 | Intron | XP_016874481.1 | ||||
XM_017018993.1 | Intron | XP_016874482.1 | ||||
XM_017018994.1 | Intron | XP_016874483.1 |
C12orf40 - chromosome 12 open reading frame 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031748.3 | Intron | NP_001026918.2 | ||||
NM_001319247.1 | Intron | NP_001306176.1 | ||||
XM_005268806.3 | Intron | XP_005268863.1 | ||||
XM_011538231.2 | Intron | XP_011536533.1 |