Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAGATTCTCCACTTTTCCGGGT[A/G]TAGATGGAAATTTCCCTTATAAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607359 MIM: 609722 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8orf58 - chromosome 8 open reading frame 58 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCAR2 - cell cycle and apoptosis regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021174.5 | Intron | NP_066997.3 | ||||
XM_011544603.1 | Intron | XP_011542905.1 | ||||
XM_011544604.1 | Intron | XP_011542906.1 | ||||
XM_017013717.1 | Intron | XP_016869206.1 |
LOC107986876 - proline-rich proteoglycan 2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017014122.1 | Intron | XP_016869611.1 |
PDLIM2 - PDZ and LIM domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |