Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTTGAGCTCCTCCAAAACCCCA[A/T]GTGACTTGTGGAAATCCCAGAAGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNRC18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TNRC18 - trinucleotide repeat containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080495.2 | Intron | NP_001073964.2 | ||||
XM_017012728.1 | Intron | XP_016868217.1 | ||||
XM_017012729.1 | Intron | XP_016868218.1 | ||||
XM_017012730.1 | Intron | XP_016868219.1 | ||||
XM_017012731.1 | Intron | XP_016868220.1 | ||||
XM_017012732.1 | Intron | XP_016868221.1 | ||||
XM_017012733.1 | Intron | XP_016868222.1 | ||||
XM_017012734.1 | Intron | XP_016868223.1 | ||||
XM_017012735.1 | Intron | XP_016868224.1 | ||||
XM_017012736.1 | Intron | XP_016868225.1 | ||||
XM_017012737.1 | Intron | XP_016868226.1 | ||||
XM_017012738.1 | Intron | XP_016868227.1 | ||||
XM_017012739.1 | Intron | XP_016868228.1 | ||||
XM_017012740.1 | Intron | XP_016868229.1 |
Set Membership: |
HapMap |