Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTTCTTCCCTGATTCCACCTG[C/T]AAATACAAACGTTATCTCTTCTCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615497 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIEF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIEF1 - mitochondrial elongation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304564.1 | Intron | NP_001291493.1 | ||||
NM_019008.5 | Intron | NP_061881.2 | ||||
XM_011530236.1 | Intron | XP_011528538.1 | ||||
XM_011530237.1 | Intron | XP_011528539.1 | ||||
XM_011530238.1 | Intron | XP_011528540.1 | ||||
XM_017028838.1 | Intron | XP_016884327.1 |