Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGGTGTGTTTTCCTCTCGGAAA[C/G]GGAATTTGGTAGCTTTGACCAATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604151 | ||||||||||||||||||||
Literature Links: |
UBE2E3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2E3 - ubiquitin conjugating enzyme E2 E3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278554.1 | Intron | NP_001265483.1 | ||||
NM_001278555.1 | Intron | NP_001265484.1 | ||||
NM_006357.3 | Intron | NP_006348.1 | ||||
NM_182678.2 | Intron | NP_872619.1 |