Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTGTCTGTAACAGTGGTTTCCA[C/T]AAAGACTTCAGTCTCATGGCATTGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601460 | |||||||||||||||||||||||
Literature Links: |
C3orf36 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
C3orf36 - chromosome 3 open reading frame 36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLCO2A1 - solute carrier organic anion transporter family member 2A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005630.2 | 3912 | UTR 3 | NP_005621.2 | |||
XM_017007075.1 | 3912 | UTR 3 | XP_016862564.1 | |||
XM_017007076.1 | 3912 | Intron | XP_016862565.1 | |||
XM_017007077.1 | 3912 | Intron | XP_016862566.1 |