Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGAAGCAGGAGGCGCGAGCCGC[C/G]CCAAAGTCTGCAACGGCCTCAAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606544 MIM: 612497 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFM2 - G elongation factor mitochondrial 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281302.1 | 17 | UTR 5 | NP_001268231.1 | |||
NM_032380.4 | 17 | UTR 5 | NP_115756.2 | |||
NM_170681.2 | 17 | UTR 5 | NP_733781.1 | |||
NM_170691.2 | 17 | UTR 5 | NP_733792.1 | |||
XM_011543691.2 | 17 | UTR 5 | XP_011541993.1 | |||
XM_017009986.1 | 17 | Intron | XP_016865475.1 |
NSA2 - NSA2, ribosome biogenesis homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271665.1 | 17 | UTR 5 | NP_001258594.1 | |||
NM_014886.4 | 17 | UTR 5 | NP_055701.1 | |||
XM_011543098.1 | 17 | Intron | XP_011541400.1 | |||
XM_017008952.1 | 17 | Intron | XP_016864441.1 | |||
XM_017008953.1 | 17 | Intron | XP_016864442.1 |