Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCAGCCATCTGCCTCCTCAATG[A/G]GTAAATTATCCCCTCTCCCCAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGBD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGBD1 - piggyBac transposable element derived 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184743.1 | Intron | NP_001171672.1 | ||||
NM_032507.3 | Intron | NP_115896.1 | ||||
XM_006715228.2 | Intron | XP_006715291.1 | ||||
XM_011514942.2 | Intron | XP_011513244.1 | ||||
XM_011514943.2 | Intron | XP_011513245.1 | ||||
XM_011514944.2 | Intron | XP_011513246.1 | ||||
XM_017011361.1 | Intron | XP_016866850.1 | ||||
XM_017011362.1 | Intron | XP_016866851.1 | ||||
XM_017011363.1 | Intron | XP_016866852.1 |
ZSCAN26 - zinc finger and SCAN domain containing 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |