Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTAGGAGTTTTTAAACATCTGAT[A/G]TAATAGCTTTTTTAAAAGGAATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928123 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928123 - uncharacterized LOC101928123 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAPLN - papilin, proteoglycan like sulfated glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173462.3 | Intron | NP_775733.3 | ||||
XM_011537290.2 | Intron | XP_011535592.1 | ||||
XM_011537291.2 | Intron | XP_011535593.1 | ||||
XM_011537292.2 | Intron | XP_011535594.1 | ||||
XM_011537293.2 | Intron | XP_011535595.1 | ||||
XM_011537294.2 | Intron | XP_011535596.1 | ||||
XM_011537295.2 | Intron | XP_011535597.1 | ||||
XM_011537296.2 | Intron | XP_011535598.1 | ||||
XM_017021749.1 | Intron | XP_016877238.1 | ||||
XM_017021750.1 | Intron | XP_016877239.1 |