Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGGCCCCGTGTCCCTGTCCCCCC[A/G]TGGCAGAAGTACTTAAGCTTAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THRA - thyroid hormone receptor, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190918.1 | Intron | NP_001177847.1 | ||||
NM_001190919.1 | Intron | NP_001177848.1 | ||||
NM_003250.5 | Intron | NP_003241.2 | ||||
NM_199334.3 | Intron | NP_955366.1 |