Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610561 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS53 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRSS53 - protease, serine 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF646 - zinc finger protein 646 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014699.3 | 472 | Intron | NP_055514.3 | |||
XM_005255710.3 | 472 | UTR 5 | XP_005255767.1 | |||
XM_005255711.3 | 472 | UTR 5 | XP_005255768.1 | |||
XM_005255712.3 | 472 | UTR 5 | XP_005255769.1 | |||
XM_011545990.2 | 472 | Intron | XP_011544292.1 |
ZNF668 - zinc finger protein 668 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172668.1 | 472 | Intron | NP_001166139.1 | |||
NM_001172669.1 | 472 | Intron | NP_001166140.1 | |||
NM_001172670.1 | 472 | Intron | NP_001166141.1 | |||
NM_024706.4 | 472 | UTR 5 | NP_078982.3 |
Set Membership: |
HapMap |