Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGCAGCTGGATGAAGTGAAGG[A/G]CACCTAAGCCAAGGGCTTTCTCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDSL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDSL - serine dehydratase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304993.1 | Intron | NP_001291922.1 | ||||
NM_138432.3 | Intron | NP_612441.1 | ||||
XM_005253831.4 | Intron | XP_005253888.1 | ||||
XM_011537846.2 | Intron | XP_011536148.1 | ||||
XM_017018763.1 | Intron | XP_016874252.1 |