Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCTCTGCATTCTCTCTAGGCCA[C/T]CTGCCCCATCCTGAGTTCACTTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300132 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRO - trophinin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039705.2 | Intron | NP_001034794.1 | ||||
NM_001271183.1 | Intron | NP_001258112.1 | ||||
NM_001271184.1 | Intron | NP_001258113.1 | ||||
NM_016157.3 | Intron | NP_057241.2 | ||||
NM_177556.2 | Intron | NP_808224.1 | ||||
NM_177557.2 | Intron | NP_808225.1 | ||||
XM_006724600.2 | Intron | XP_006724663.1 | ||||
XM_011530808.1 | Intron | XP_011529110.1 | ||||
XM_011530809.1 | Intron | XP_011529111.1 | ||||
XM_011530811.2 | Intron | XP_011529113.1 | ||||
XM_011530812.1 | Intron | XP_011529114.1 | ||||
XM_011530813.1 | Intron | XP_011529115.1 | ||||
XM_011530814.1 | Intron | XP_011529116.1 | ||||
XM_017029767.1 | Intron | XP_016885256.1 | ||||
XM_017029768.1 | Intron | XP_016885257.1 | ||||
XM_017029769.1 | Intron | XP_016885258.1 | ||||
XM_017029770.1 | Intron | XP_016885259.1 | ||||
XM_017029771.1 | Intron | XP_016885260.1 | ||||
XM_017029772.1 | Intron | XP_016885261.1 | ||||
XM_017029773.1 | Intron | XP_016885262.1 |