Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCAAAATACCTAATCTTTCAGT[G/T]TTAATACCTTTCAAATTTCAATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601232 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3CG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3CG - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282426.1 | Intron | NP_001269355.1 | ||||
NM_001282427.1 | Intron | NP_001269356.1 | ||||
NM_002649.3 | Intron | NP_002640.2 | ||||
XM_005250443.3 | Intron | XP_005250500.1 | ||||
XM_011516316.1 | Intron | XP_011514618.1 | ||||
XM_011516317.1 | Intron | XP_011514619.1 | ||||
XM_017012328.1 | Intron | XP_016867817.1 |