Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTGCCTAACTCTATTTATGTTT[C/T]TTTACTATCATAACACTTCATGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607222 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD16 - ankyrin repeat domain 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO18 - F-box protein, helicase, 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258452.1 | 393 | Intron | NP_001245381.1 | |||
NM_001258453.1 | 393 | Intron | NP_001245382.1 | |||
NM_032807.4 | 393 | Intron | NP_116196.3 | |||
NM_178150.2 | 393 | Intron | NP_835363.1 | |||
XM_011519748.2 | 393 | Intron | XP_011518050.1 | |||
XM_017016842.1 | 393 | UTR 5 | XP_016872331.1 | |||
XM_017016843.1 | 393 | Intron | XP_016872332.1 | |||
XM_017016844.1 | 393 | Intron | XP_016872333.1 | |||
XM_017016845.1 | 393 | Intron | XP_016872334.1 | |||
XM_017016846.1 | 393 | Intron | XP_016872335.1 |