Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606597 | ||||||||||||||||||||
Literature Links: |
CCDC140 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC140 - coiled-coil domain containing 140 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153038.1 | 226 | Intron | NP_694583.1 |
PAX3 - paired box 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000438.5 | 226 | UTR 5 | NP_000429.2 | |||
NM_001127366.2 | 226 | UTR 5 | NP_001120838.1 | |||
NM_013942.4 | 226 | UTR 5 | NP_039230.1 | |||
NM_181457.3 | 226 | UTR 5 | NP_852122.1 | |||
NM_181458.3 | 226 | UTR 5 | NP_852123.1 | |||
NM_181459.3 | 226 | UTR 5 | NP_852124.1 | |||
NM_181460.3 | 226 | UTR 5 | NP_852125.1 | |||
NM_181461.3 | 226 | UTR 5 | NP_852126.1 |